2'-Deoxyinosine-5'-monophosphate disodium salt 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Sodium ((2R,3S,5R)-3-hydroxy-5-(6-oxo-1H-purin-9(6H)-yl)tetrahydrofuran-2-yl)methyl phosphate
J64271.03
J64271.03EA
80.9
EUR
J64271.03
2'-Deoxyinosine-5'-monophosphate disodium salt 99%
2'-Deoxyinosine-5'-monophosphate disodium salt
Purity: 99%
CAS No.: 14999-52-1
Molecular Formula: C10H11N4Na2O7P
Formula Weight: 376.17
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-whgite to white
MDL No.: MFCD00134874
Formel:
C₁₀H₁₁N₄Na₂O₇P Molecular Weight: 376,17 g/mol |
MDL:
MFCD00134874 CAS-Nummer: 14999-52-1 EINECS: 239-090-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...