Coomassie® Brillantblau R 250
Lieferant: Thermo Fisher Scientific
Synonyme:
Brillantblau R, Hydrogen[4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylen]cyclohexa-2,5-dien-1-yliden](ethyl)(m-sulfonatobenzyl)ammonium Mononatriumsalz, Coomassie Blau R 250, Xylenbrillantcyanin, Serva Blue R, Brillantindocyanin 6B, Acid Blue 83
J64297.18
J64297.18EA
87.5
EUR
J64297.18
Coomassie® Brillantblau R 250
Coomassie® Brillantblau R 250
Formel:
C₄₅H₄₄N₃NaO₇S₂ Molecular Weight: 825.98 g/mol Schmelzpunkt: 174…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041762 CAS-Nummer: 6104-59-2 EINECS: 228-060-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...