Cresylviolett (Azetat)

Lieferant: Thermo Scientific

Synonyme: Cresyl echt violet, Cresyl violet, Cresyl fast violet, 5,9-Diaminobenzo[a]phenoxazin-7-ium acetate, 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt, Cresyl Violet acetate, Gentian violet

J64318.09
J64318.09EA 127 EUR
J64318.09
Cresylviolett (Azetat)
Cresylviolett (Azetat)

CAS No.: 10510-54-0 Molecular Formula: C18H15N3O3 Formula Weight: 321.33 Physical Form: Powder Appearance: Dark green to black Melting Point: 140-143° MDL No.: MFCD00013151


Applications: Stain for nerve cells and tissue

Formel: C₁₈H₁₅N₃O₃
Molecular Weight: 321,33 g/mol
Schmelzpunkt: 140…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013151
CAS-Nummer: 10510-54-0
EINECS: 234-043-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR