Citronensäure ≥99,5% für die Elektrophorese
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.5+%
CAS No.: 77-92-9
Molecular Formula: C6H8O7
Formula Weight: 192.13
Physical Form: Powder
Density: 1.665
Appearance: White
Odour: Odourless
pH: 1,6
Melting Point: 153-154°
Beilstein Reference: 782061
Merck Reference: 14,2326
MDL No.: MFCD00011669
Formel:
HOC(COOH)(CH₂COOH)₂ Siedepunkt: 310 °C (1013 hPa) Schmelzpunkt: 153…154 °C Dichte: 1,665 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011669 CAS-Nummer: 77-92-9 EINECS: 201-069-1 REACH: 01-2119457026-42-0000 Merck Index: 14,2326 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...