DNS Natriumsalz (Desoxyribonukleinsäure Natriumsalz) (aus Kalbsthymus)
Lieferant: Thermo Fisher Scientific
Deoxyribonucleic acid sodium salt, is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard to determine DNA content in gels and for DNA quantification by fluorescent assay.
CAS No.: 91080-16-9
Storage Temperature: 2°C to 8°C
Physical Form: Lyophilized powder
Appearance: White
MDL No.: MFCD00151698
Applications: Suitable for use as a substrate in DNase assays.
Lagertemperatur:
Kühlschrank |
MDL:
MFCD00151698 CAS-Nummer: 9007-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...