2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid
Lieferant: Thermo Scientific
J64414.MA
J64414.MAEA
170
EUR
J64414.MA
2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid
2-(4,5,6,7-Tetraiodo-1,3-dioxoisoindolin-2-yl)acetic acid
CAS No.: 19231-60-8
Molecular Formula: C10H3I4NO4
Formula Weight: 708.75
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Crystalline
MDL No.: MFCD05023065
Formel:
C₁₀H₃I₄NO₄ Molecular Weight: 708,75 g/mol |
MDL:
MFCD05023065 CAS-Nummer: 19231-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...