Sirtinol
Lieferant: Thermo Fisher Scientific
|
Achtung
|
J64486.MA
J64486.MAEA
155
EUR
J64486.MA
Sirtinol
Sirtinol
CAS No.: 410536-97-9
Molecular Formula: C26H22N2O2
Formula Weight: 394.46
Storage Temperature: -30°C to -10°C
Physical Form: Crystals
Appearance: Off-white to yellow
Solubility: Soluble in water, DMSO and methanol
MDL No.: MFCD00810186
Applications: Sirtinol is a cell permeable specific inhibitor of sirtuin NAD-dependant deacetylases
Formel:
C₂₆H₂₂N₂O₂ |
MDL:
MFCD00810186 CAS-Nummer: 410536-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...