Z-Val-Val-Nle diazomethyl ketone
Lieferant: Thermo Fisher Scientific
J64544.MF
J64544.MFEA
365
EUR
J64544.MF
Z-Val-Val-Nle diazomethyl ketone
Z-Val-Val-Nle diazomethyl ketone
CAS No.: 155026-49-6
Molecular Formula: C25H37N5O5
Formula Weight: 487.60
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00672348
Applications: Inactivates thiol proteinases; an irreversible cathepsin inhibitor
Formel:
C₂₅H₃₇N₅O₅ |
MDL:
MFCD00672348 CAS-Nummer: 155026-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...