2',3'-Didesoxyadenosin ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
|
Purity: 98%
CAS No.: 4097-22-7
Molecular Formula: C10H13N5O2
Formula Weight: 235.24
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Melting Point: 181-184°
Beilstein Reference: 619924
Merck Reference: 14,3101
MDL No.: MFCD00010534
Formel:
C₁₀H₁₃N₅O₂ Schmelzpunkt: 181…184 °C |
MDL:
MFCD00010534 CAS-Nummer: 4097-22-7 EINECS: 223-853-2 Merck Index: 14,3101 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...