N-Phthalyl-L-tryptophan ≥99%

Lieferant: Thermo Scientific

Synonyme: (S)-2-(1,3-Dioxoisoindolin-2-yl)-3-(1H-indol-3-yl)propanoic acid, RG 108, RG108

J64630.MA
J64630.MAEA 131 EUR
J64630.MA
N-Phthalyl-L-tryptophan ≥99%
N-Phthalyl-L-tryptophan

A cell-permeable, specific DNA methyltransferases inhibitor.


Yellow solid.

Formel: C₁₉H₁₄N₂O₄
Molecular Weight: 334,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08705332
CAS-Nummer: 48208-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR