1,4-Dithioerythrit 99% für die Elektrophorese
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 6892-68-8
Molecular Formula: C4H10O2S2
Formula Weight: 154.25
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: White
Odour: Stench
Melting Point: 82-84°
Beilstein Reference: 1719756
UN No.: 3335
MDL No.: MFCD00063750
Formel:
C₄H₁₀O₂S₂ Siedepunkt: 286 °C (1013 hPa) Schmelzpunkt: 82…84 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00063750 CAS-Nummer: 6892-68-8 EINECS: 229-998-8 UN: 3335 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...