Glycylphenylalanine-2-naphthylamide

Lieferant: Thermo Scientific
Achtung

Synonyme: H-Gly-Phe-βNA

J64718.MC
J64718.MCEA 80.2 EUR
J64718.MC
Glycylphenylalanine-2-naphthylamide
Glycylphenylalanine-2-naphthylamide

CAS No.: 21438-66-4 Molecular Formula: C21H21N3O2 Formula Weight: 347.41 Storage Temperature: -30°C to -10°C Physical Form: Crystalline MDL No.: MFCD00021597


Applications: Substrate for cathepsin C

Formel: C₂₁H₂₁N₃O₂
Molecular Weight: 347,418 g/mol
MDL: MFCD00021597
CAS-Nummer: 21438-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR