8-Brom-2'-desoxyguanosin 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Amino-8-bromo-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one
J64723.03
J64723.03EA
111
EUR
J64723.03
8-Brom-2'-desoxyguanosin 99%
8-Brom-2'-desoxyguanosin
Purity: 99%
CAS No.: 13389-03-2
Molecular Formula: C10H12BrN5O4
Formula Weight: 346.14
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point:
Formel:
C₁₀H₁₂BrN₅O₄ Molecular Weight: 346.14 g/mol Schmelzpunkt: >250 °C (Zers.) |
MDL:
MFCD01630971 CAS-Nummer: 13389-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...