Glu-NH-med
Lieferant: Thermo Scientific
Synonyme:
H-Glu-AMC
J64847.MF
J64847.MFEA
114
EUR
J64847.MF
Glu-NH-med
Glu-NH-med
CAS No.: 98516-76-8
Molecular Formula: C15H16N2O5
Formula Weight: 304.30
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00051217
Applications: A fluorogenic substrate for aminopeptidase a, producing a blue fluorescent solution upon cleavage.
Formel:
C₁₅H₁₆N₂O₅ Molecular Weight: 304,302 g/mol |
MDL:
MFCD00051217 CAS-Nummer: 98516-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...