6-Chloropurine 2'-deoxyriboside 97%

Lieferant: Thermo Fisher Scientific
Achtung

J64871.03
J64871.03EA 300 EUR
J64871.03
6-Chloropurine 2'-deoxyriboside 97%
6-Chloropurine 2'-deoxyriboside

Purity: 97% CAS No.: 4594-45-0 Molecular Formula: C10H11ClN4O3 Formula Weight: 270.67 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to off-white MDL No.: MFCD00083282

Formel: C₁₀H₁₁ClN₄O₃
Molecular Weight: 270.673 g/mol
MDL: MFCD00083282
CAS-Nummer: 4594-45-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR