5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine 98%
Lieferant: Thermo Scientific
J64880.03
J64880.03EA
40.7
EUR
J64880.03
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine 98%
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine
Purity: 98%
Molecular Formula: C31H32N2O8
Formula Weight: 560.59
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Soluble in acetonitrile
MDL No.: MFCD00274123