5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine 98%
Lieferant: Thermo Scientific
J64880.03
J64880.03EA 40.7 EUR
J64880.03
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine 98%
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine
Purity: 98% Molecular Formula: C31H32N2O8 Formula Weight: 560.59 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale yellow Solubility: Soluble in acetonitrile MDL No.: MFCD00274123