2'-O-Methyluridin 99%

Lieferant: Thermo Scientific

Synonyme: 1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 2′-O-Methyluridine

J64897.03
J64897.03EA 30 EUR
J64897.03
2'-O-Methyluridin 99%
2'-O-Methyluridine

Purity: 99% CAS No.: 2140-76-3 Molecular Formula: C10H14N2O6 Formula Weight: 258.23 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00056054

Formel: C₁₀H₁₄N₂O₆
Molecular Weight: 258,23 g/mol
MDL: MFCD00056054
CAS-Nummer: 2140-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR