N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride

Lieferant: Thermo Scientific

Synonyme: Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride, Bz-Arg-pNA·HCl, Bz-Arg-PNA.HCl, N-α-Benzoyl-L-Arginine p-Nitroanilide Hydrochloride, (S)-N-(5-Guanidino-1-((4-nitrophenyl)amino)-1-oxopentan-2-yl)benzamide hydrochloride

J64915.ME
J64915.MEEA 274 EUR
J64915.ME
N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride

CAS No.: 21653-40-7 Molecular Formula: C19H22N6O4.HCl Formula Weight: 434.90 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to yellow Melting Point: 223° Solubility: Soluble in DMSO at 50mg/ml Beilstein Reference: 4081878 MDL No.: MFCD00063682


Applications: Chromogenic trypsin substrate. Substrate for proteolytic enzymes

Formel: C₁₉H₂₃ClN₆O₄
Molecular Weight: 434,881 g/mol
Schmelzpunkt: 223 °C
MDL: MFCD00063682
CAS-Nummer: 21653-40-7
EINECS: 244-505-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR