2'-Deoxythymidine-5'-diphosphate trisodium salt 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
J64938.MB
J64938.MBEA
165
EUR
J64938.MB
2'-Deoxythymidine-5'-diphosphate trisodium salt 97%
2'-Deoxythymidine-5'-diphosphate trisodium salt
Purity: 97%
CAS No.: 95648-78-5
Molecular Formula: C10H13N2Na3O11P2
Formula Weight: 468.13
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00084748
Formel:
C₃₀H₄₅N₆Na₃O₃₃P₆ Molecular Weight: 1272.511 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00084748 CAS-Nummer: 95648-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...