MIF antagonist, ISO-1
Lieferant: Thermo Scientific
Synonyme:
Methyl 2-(3-(4-hydroxyphenyl)-4,5-dihydroisoxazol-5-yl)acetate, ISO-1
J64947.MB
J64947.MBEA
332
EUR
J64947.MB
MIF antagonist, ISO-1
MIF antagonist, ISO-1
CAS No.: 478336-92-4
Molecular Formula: C12H13NO4
Formula Weight: 235.24
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Pale yellow
MDL No.: MFCD22376813
Applications: A cell-permeable isoxazoline compound
Formel:
C₁₂H₁₃NO₄ Molecular Weight: 235,2 g/mol |
MDL:
MFCD22376813 CAS-Nummer: 478336-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...