L-Leucine(4-methyl-7-coumarinylamide), Hyrdrat
Lieferant: Thermo Scientific
Synonyme:
H-Leu-AMC
J64951.MD
J64951.MDEA
522
EUR
J64951.MD
L-Leucine(4-methyl-7-coumarinylamide), Hyrdrat
L-Leucine(4-methyl-7-coumarinylamide)
CAS No.: 66447-31-2
Molecular Formula: C16H20N2O3.xH2O
Formula Weight: 288.37 (anhy)
Physical Form: Powder
Appearance: White to pale yellow
MDL No.: MFCD04966490
Applications: A sensitive fluorogenic substrate for leucine aminopeptidase and aminopeptidase M
Formel:
C₁₆H₂₀N₂O₃·xH₂O Molecular Weight: 288,347 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04966490 CAS-Nummer: 66447-31-2 EINECS: 266-363-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...