N6-Methyl-2'-deoxyadenosine 99%
Lieferant: Thermo Scientific
Synonyme:
(2R,3S,5R)-2-(Hydroxymethyl)-5-(6-(methylamino)-9H-purin-9-yl)tetrahydrofuran-3-ol, 2'-Deoxy-N6-methyladenosine
J64961.03
J64961.03EA
391
EUR
J64961.03
N6-Methyl-2'-deoxyadenosine 99%
N6-Methyl-2'-deoxyadenosine
Purity: 99%
CAS No.: 2002-35-9
Molecular Formula: C11H15N5O3
Formula Weight: 265.27
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water
MDL No.: MFCD00055999
Formel:
C₁₁H₁₅N₅O₃ Molecular Weight: 265,273 g/mol |
MDL:
MFCD00055999 CAS-Nummer: 2002-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...