Azadibenzocyclooctyne-PEG, M.W. 5 kDa

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: ADIBO-PEG

J64977.MCEA 197 EUR
J64977.MC
Azadibenzocyclooctyne-PEG, M.W. 5 kDa
Azadibenzocyclooctyne-PEG

Azadibenzocyclooctyne-PEG is used for copper-free click chemistry reactions.


Appearance: White to grey solid


Bitte beachten Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR