2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride
Lieferant: Thermo Scientific
J64981.MA
J64981.MAEA
126
EUR
J64981.MA
2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride
2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride
CAS No.: 418788-90-6
Molecular Formula: C11H15Cl2NO2.HCl
Formula Weight: 300.61
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water at ~25 mg/ml. Also soluble in ethanol, DMSO or DMF at ~20mg/ml. Soluble in PBS, pH 7.2 at ~10 mg/ml.
MDL No.: MFCD06410993
Formel:
C₁₁H₁₅Cl₂NO₂·HCl Molecular Weight: 300,61 g/mol |
MDL:
MFCD06410993 CAS-Nummer: 418788-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...