N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine 98%
Lieferant: Thermo Scientific
J65098.03
J65098.03EA
55.5
EUR
J65098.03
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine 98%
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine
Purity: 98%
Molecular Formula: C39H37N5O7
Formula Weight: 687.74
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Soluble in chloroform at 50mg/ml
MDL No.: MFCD08704196