N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine 98%
Lieferant: Thermo Scientific
J65098.03
J65098.03EA 55.5 EUR
J65098.03
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine 98%
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine
Purity: 98% Molecular Formula: C39H37N5O7 Formula Weight: 687.74 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale yellow Solubility: Soluble in chloroform at 50mg/ml MDL No.: MFCD08704196