5'-O-(4,4'-Dimethoxytrityl)-N-2-isobutyryl-2'-O-methylguanosine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
Lieferant: Thermo Scientific
|
Achtung
|
J65100.03
J65100.03EA
64.4
EUR
J65100.03
5'-O-(4,4'-Dimethoxytrityl)-N-2-isobutyryl-2'-O-methylguanosine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
5'-O-(4,4'-Dimethoxytrityl)-N-2-isobutyryl-2'-O-methylguanosine-3'-(2-cyanoethyl diisopropylphosphoramidite)
Purity: 98%
Molecular Formula: C45H56N7O9P
Formula Weight: 869.94
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00792625
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...