IKK-2 Inhibitor V

Lieferant: Thermo Scientific
Achtung

Synonyme: IMD-0354, N-(3,5-Bis-trifluoromethylphenyl)-5-chloro-2-hydroxybenzamide, N-(3,5-Bis(trifluoromethyl)phenyl)-5-chloro-2-hydroxybenzamide

J65132.MA
J65132.MAEA 131 EUR
J65132.MA
IKK-2 Inhibitor V
IKK-2

A selective IKK-2 inhibitor.


  • Light Sensitive


White powder

Formel: C₁₅H₈ClF₆NO₂
Molecular Weight: 383,67 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00218820
CAS-Nummer: 978-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR