5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyinosine 98%

Lieferant: Thermo Scientific

J65136.03
J65136.03EA 45.9 EUR
J65136.03
5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyinosine 98%
5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyinosine

Purity: 98% CAS No.: 93778-57-5 Molecular Formula: C31H30N4O6 Formula Weight: 554.59 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to pale yellow MDL No.: MFCD00080301

Formel: C₃₁H₃₀N₄O₆
Molecular Weight: 554,59 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00080301
CAS-Nummer: 93778-57-5
EINECS: 298-195-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR