D(+)-Saccharose Molecular_Biology
Lieferant: Thermo Fisher Scientific
Synonyme:
Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker
J65148.36
J65148.A1
J65148.36EA
44.6
EUR
J65148.36
J65148.A1
D(+)-Saccharose Molecular_Biology
D(+)-Saccharose
CAS No.: 57-50-1
Molecular Formula: C12H22O11
Formula Weight: 342.30
Physical Form: Crystalline solid
Density: 1.58
Appearance: White
Odour: Odourless
Melting Point: 185-187°
Solubility: Soluble in water
Beilstein Reference: 90825
MDL No.: MFCD00006626
For protein purification by sucrose density gradients
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342.3 g/mol Schmelzpunkt: 185…187 °C Dichte: 1.58 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006626 CAS-Nummer: 57-50-1 EINECS: 200-334-9 Merck Index: 13,08966 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...