AM 580

Lieferant: Thermo Scientific

Synonyme: NSC 608001, CD336, 4-​​[[(5,​​6,​​7,​​8-​​tetrahydro-​​5,​​5,​​8,​​8-​​tetramethyl-​​2-​​naphthalenyl)​carbonyl]​amino]​-​benzoic acid, AM580, 4-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-carboxamido)benzoic acid, RO 40-6055

J65155.MA
J65155.MAEA 128 EUR
J65155.MA
AM 580
AM 580

A retinoid derivative used for gene expression studies of leukemia-retinoic acid receptor (PHL-RAR) cell line.


  • Soluble in DMSO (20mg/ml) or ethanol (10mg/ml).


White crystalline solid.

Formel: C₂₂H₂₅NO₃
Molecular Weight: 351,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00673916
CAS-Nummer: 102121-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR