3,4-Dichloroisocoumarin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3,4-Dichloro-1H-isochromen-1-one
Useful in a rapid staining method for identification of macrophages; counts by this method were confirmed by the more complex morphological criteria, by phagocytosis, and by the presence of Fc receptors. It is an inhibitor of Chymotrypsin and Trypsin. ,4 Dichloroisocoumarin also inhibits granzymes A, B, and H, cathepsin G, neutrophil elastase, proteinase and blocks apoptotic internucleosomal DNA cleavage in thymocytes without the involvement of endonucleases.
Reversible inhibitor of serine proteases
Formel:
C₉H₄Cl₂O₂ Molecular Weight: 215,029 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00036960 CAS-Nummer: 51050-59-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...