2'-Fluoro-2'-deoxyinosine 99%
Lieferant: Thermo Fisher Scientific
J65208.03
J65208.03EA
377
EUR
J65208.03
2'-Fluoro-2'-deoxyinosine 99%
2'-Fluoro-2'-deoxyinosine
Purity: 99%
CAS No.: 80049-87-2
Molecular Formula: C10H11FN4O4
Formula Weight: 270.22
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD09750860
Formel:
C₁₀H₁₁FN₄O₄ |
MDL:
MFCD09750860 CAS-Nummer: 80049-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...