Tyrphostin C15
Lieferant: Thermo Fisher Scientific
Synonyme:
AG-825, 3-(3-((Benzo[d]thiazol-2-ylthio)methyl)-4-hydroxy-5-methoxyphenyl)-2-cyanoacrylamide
J65227.MA
J65227.MAEA
206
EUR
J65227.MA
Tyrphostin C15
Tyrphostin C15
CAS No.: 149092-50-2
Molecular Formula: C19H15N3O3S2
Formula Weight: 397.47
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Yellow to orange
MDL No.: MFCD01074971
Applications: A selective ErbB2 inhibitor
Formel:
C₁₉H₁₅N₃O₃S₂ Molecular Weight: 397.5 g/mol |
MDL:
MFCD01074971 CAS-Nummer: 149092-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...