SANT-1

Lieferant: Thermo Scientific

Synonyme: N-(4-Benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanimine

J65294.MA
J65294.MAEA 156 EUR
J65294.MA
SANT-1
SANT-1

Potent, cell-permeable inhibitor of Sonic hedgehog (Shh) signaling; high affinity antagonist of smoothened activity.


  • Moisture and Light Sensitive


Pale yellow Solid


Soluble in DMSO with warming

Formel: C₂₃H₂₇N₅
Molecular Weight: 373,5 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01827306
CAS-Nummer: 304909-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR