8-Bromo-2'-deoxyadenosine 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
J65357.03
J65357.03EA
136
EUR
J65357.03
8-Bromo-2'-deoxyadenosine 99%
8-Bromo-2'-deoxyadenosine
Purity: 99%
CAS No.: 14985-44-5
Molecular Formula: C10H12FN5O3
Formula Weight: 284.25
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to yellow
Solubility: Soluble in DMSO or DMF
MDL No.: MFCD01630970
Formel:
C₁₀H₁₂BrN₅O₃ Molecular Weight: 330.142 g/mol |
MDL:
MFCD01630970 CAS-Nummer: 14985-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...