2,4,6-Trimethyl-N-(2-(trifluormethyl)phenyl)benzolsulfonamid
Lieferant: Thermo Fisher Scientific
J65391.MA
J65391.MAEA
95.9
EUR
J65391.MA
2,4,6-Trimethyl-N-(2-(trifluormethyl)phenyl)benzolsulfonamid
2,4,6-Trimethyl-N-(2-(trifluormethyl)phenyl)benzolsulfonamid
CAS No.: 313981-55-4
Molecular Formula: C16H16F3NO2S
Formula Weight: 343.36
Physical Form: Powder
Appearance: White
An inactive analog of m-3M3FBS. Used as a negative control.
Formel:
C₁₆H₁₆F₃NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00784362 CAS-Nummer: 313981-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...