N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine 98%
Lieferant: Thermo Scientific
J65402.03
J65402.03EA
129
EUR
J65402.03
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine 98%
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine
Purity: 98%
Molecular Formula: C38H34FN5O6
Formula Weight: 675.70
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
MDL No.: MFCD15145191
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...