2'-Fluoro-2'-deoxyadenosine 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 2′-Deoxy-2′-fluoroadenosine, (2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)tetrahydrofuran-3-ol, 2'-Deoxy-2'-fluoroadenosine

J65441.03
J65441.03EA 114 EUR
J65441.03
2'-Fluoro-2'-deoxyadenosine 99%
2'-Fluoro-2'-deoxyadenosine

Purity: 99% CAS No.: 64183-27-3 Molecular Formula: C10H12FN5O3 Formula Weight: 269.23 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD09750859

Formel: C₁₀H₁₂FN₅O₃
Molecular Weight: 269.236 g/mol
MDL: MFCD09750859
CAS-Nummer: 64183-27-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR