5-Bromo-2′-deoxycytidine 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-Amino-5-bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

J65456.03
J65456.03EA 65.8 EUR
J65456.03
5-Bromo-2′-deoxycytidine 99%
5-Bromo-2′-deoxycytidine

Purity: 99% CAS No.: 1022-79-3 Molecular Formula: C9H11BrN2O5 Formula Weight: 307.10 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale yellow MDL No.: MFCD00047496

Formel: C₉H₁₂BrN₃O₄
Molecular Weight: 306.116 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00047496
CAS-Nummer: 1022-79-3
EINECS: 213-824-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR