5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
Lieferant: Thermo Scientific
Achtung
J65508.03
J65508.03EA 37.1 EUR
J65508.03
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine-3'-(2-cyanoethyl diisopropylphosphoramidite)
Purity: 98% Molecular Formula: C40H49N4O9P Formula Weight: 760.81 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00792626