Arabinofuranosyl-2-fluoroadenine 98%
Lieferant: Thermo Fisher Scientific
Purity: 98%
CAS No.: 21679-14-1
Molecular Formula: C10H12FN5O4
Formula Weight: 285.23
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Soluble in DMF, DMSO, methanol or ethanol. Sparingly soluble in water
Beilstein Reference: 1225932
MDL No.: MFCD00132942
Formel:
C₁₀H₁₂FN₅O₄ |
MDL:
MFCD00132942 CAS-Nummer: 21679-14-1 EINECS: 244-525-5 Merck Index: 14,4126 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...