5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine 98%

Lieferant: Thermo Scientific

Synonyme: N-(9-((2R,3R,4R,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxy-3-methoxytetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-1H-purin-2-yl)isobutyramide, 5'-O-DMT-N2-Isobutyryl-2'-O-methyl-D-guanosine

J65583.03
J65583.03EA 77.3 EUR
J65583.03
5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine 98%
5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine

Purity: 98% CAS No.: 114745-26-5 Molecular Formula: C36H39N5O8 Formula Weight: 669.72 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale yellow Solubility: Soluble in acetonitrile MDL No.: MFCD09842119

Formel: C₃₆H₃₉N₅O₈
Molecular Weight: 669,72 g/mol
MDL: MFCD09842119
CAS-Nummer: 114745-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR