L-Glycine 7-amido-4-methylcoumarin
Lieferant: Thermo Scientific
J65592.MA
J65592.MAEA
59.5
EUR
J65592.MA
L-Glycine 7-amido-4-methylcoumarin
L-Glycine 7-amido-4-methylcoumarin
Molecular Formula: C12H12N2O3
Formula Weight: 232.23
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
Solubility: Soluble in DMSO or water
MDL No.: MFCD00077122
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...