2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine 98%
Lieferant: Thermo Scientific
J65608.03
J65608.03EA
127
EUR
J65608.03
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine 98%
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine
Purity: 98%
Molecular Formula: C14H18FN5O5
Formula Weight: 355.32
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Soluble in methanol
MDL No.: MFCD15145246