2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine 98%
Lieferant: Thermo Scientific
J65608.03
J65608.03EA 127 EUR
J65608.03
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine 98%
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine
Purity: 98% Molecular Formula: C14H18FN5O5 Formula Weight: 355.32 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale yellow Solubility: Soluble in methanol MDL No.: MFCD15145246