2'-Fluoro-2'-deoxycytidine 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-1-((2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, 2'-Deoxy-2'-fluorocytidine

J65632.03
J65632.03EA 53.3 EUR
J65632.03
2'-Fluoro-2'-deoxycytidine 99%
2'-Fluoro-2'-deoxycytidine

Purity: 99% CAS No.: 10212-20-1 Molecular Formula: C9H12FN3O4 Formula Weight: 245.21 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00057445

Formel: C₉H₁₂FN₃O₄
Molecular Weight: 245,21 g/mol
MDL: MFCD00057445
CAS-Nummer: 10212-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR