GW-9662

Lieferant: Thermo Scientific
Achtung

Synonyme: GW9662, 2-Chloro-5-nitro-N-phenylbenzamide

J65634.MA
J65634.MAEA 98.8 EUR
J65634.MA
GW-9662
GW-9662

An irreversible PPAR antagonist


Off-white powder

Formel: C₁₃H₉N₂O₃Cl
Molecular Weight: 276,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01215270
CAS-Nummer: 22978-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR