N-Acetyl-L-histidine monohydrate

Lieferant: Thermo Scientific

Synonyme: Nα-Acetyl-L-histidine monohydrate, Nα-Acetyl-L-histidine, N-Acetyl-L-histidine Monohydrate, Ac-His-OH.H2O, Ac-His-OH·H2O, N-Ac-His-OH·H₂O

J65657.06
J65657.06EA 159 EUR
J65657.06
N-Acetyl-L-histidine monohydrate
N-Acetyl-L-histidine monohydrate

CAS No.: 39145-52-3 Molecular Formula: C8H11N3O3.H2O Formula Weight: 215.21 (197.09anhy) Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 163° Optical Rotation: +47°(c=1 in water) MDL No.: MFCD00149320

Formel: C₈H₁₁N₃O₃·H₂O
Molecular Weight: 215,2 g/mol
Schmelzpunkt: 163 °C
MDL: MFCD00149320
CAS-Nummer: 39145-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR