2'-Deoxycytidine-5'-monophosphate disodium salt

Lieferant: Thermo Scientific

Synonyme: Sodium ((2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3-hydroxytetrahydrofuran-2-yl)methyl phosphate, Disodium 2'-deoxy-5'-O-phosphonatocytidine

J65724.03
J65724.03EA 80.5 EUR
J65724.03
2'-Deoxycytidine-5'-monophosphate disodium salt
2'-Deoxycytidine-5'-monophosphate disodium salt

CAS No.: 13085-50-2 Molecular Formula: C9H12N3Na2O7P Formula Weight: 351.16 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00069774

Formel: C₉H₁₂N₃Na₂O₇P
Molecular Weight: 351,16 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00069774
CAS-Nummer: 13085-50-2
EINECS: 235-995-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR