Harnstoff 99%, ultrarein
Lieferant: Thermo Fisher Scientific
Synonyme:
Urea, Aminoameisensäureamid, Carbamid, Carbonyldiamid
J65769.22
J65769.36
J65769.A4
J65769.22EA
31.9
EUR
J65769.22
J65769.36
J65769.A4
Harnstoff 99%, ultrarein
Harnstoff
Purity: 99%
CAS No.: 57-13-6
Molecular Formula: CH4N2O
Formula Weight: 60.06
Physical Form: Crystalline
Density: 1.335
Appearance: White
Odour: Ammonia-like
pH: 9,2-9,5
Melting Point: 132-135°
Solubility: Soluble in water
Beilstein Reference: 635724
Merck Reference: 14,9867
MDL No.: MFCD00008022
Formel:
NH₂CONH₂ Siedepunkt: 197 °C (1013 hPa) Schmelzpunkt: 132…135 °C Dichte: 1,335 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008022 CAS-Nummer: 57-13-6 EINECS: 200-315-5 REACH: 01-2119463277-33 Merck Index: 14,9867 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...