Harnstoff 99%, ultrarein

Lieferant: Thermo Fisher Scientific

Synonyme: Urea, Aminoameisensäureamid, Carbamid, Carbonyldiamid

J65769.22 J65769.36 J65769.A4
J65769.22EA 31.9 EUR
J65769.22 J65769.36 J65769.A4
Harnstoff 99%, ultrarein
Harnstoff

Purity: 99% CAS No.: 57-13-6 Molecular Formula: CH4N2O Formula Weight: 60.06 Physical Form: Crystalline Density: 1.335 Appearance: White Odour: Ammonia-like pH: 9,2-9,5 Melting Point: 132-135° Solubility: Soluble in water Beilstein Reference: 635724 Merck Reference: 14,9867 MDL No.: MFCD00008022

Formel: NH₂CONH₂
Siedepunkt: 197 °C (1013 hPa)
Schmelzpunkt: 132…135 °C
Dichte: 1,335
Lagertemperatur: Raumtemperatur
MDL: MFCD00008022
CAS-Nummer: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 14,9867
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR